2-Nitro-3-(trifluoromethyl)aniline

Lieferant: Apollo Scientific
Achtung

PC53489-1G PC53489-250MG PC53489-5G
APOSPC53489-1GEA 219 EUR
APOSPC53489-1G APOSPC53489-250MG APOSPC53489-5G
2-Nitro-3-(trifluoromethyl)aniline
2-Nitro-3-(trifluoromethyl)aniline
Formel: C₇H₅F₃N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00042448
CAS-Nummer: 386-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR