6-Amino-7-bromo-2,2,3,3-tetrafluoro-1,4-benzodioxane
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromo-2,2,3,3-tetrafluoro-2,3-dihydro-1,4-benzodioxin-6-amine
PC53485-1G
PC53485-250MG
APOSPC53485-1GEA
627
EUR
APOSPC53485-1G
APOSPC53485-250MG
6-Amino-7-bromo-2,2,3,3-tetrafluoro-1,4-benzodioxane
6-Amino-7-bromo-2,2,3,3-tetrafluoro-1,4-benzodioxane
Formel:
C₈H₄BrF₄NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD30723347 CAS-Nummer: 254895-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...