4-Bromo-2,5-difluorobenzylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-2,5-difluorophenyl)methanamine hydrochloride

PC53452-1G PC53452-250MG
APOSPC53452-1GEA 560 EUR
APOSPC53452-1G APOSPC53452-250MG
4-Bromo-2,5-difluorobenzylamine hydrochloride
4-Bromo-2,5-difluorobenzylamine hydrochloride
Formel: C₇H₆BrF₂N·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD30497599
CAS-Nummer: 1256276-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR