4-Bromo-2-fluoro-5-nitrobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC53405-1G PC53405-250MG
APOSPC53405-1GEA 36.4 EUR
APOSPC53405-1G APOSPC53405-250MG
4-Bromo-2-fluoro-5-nitrobenzoic acid
4-Bromo-2-fluoro-5-nitrobenzoic acid
Formel: C₇H₃BrFNO₄
Molekulargewicht: 264.005 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11977380
CAS-Nummer: 355423-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR