4-Bromo-2-fluoro-5-nitrobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC53377-1G PC53377-250MG
APOSPC53377-1GEA 429 EUR
APOSPC53377-1G APOSPC53377-250MG
4-Bromo-2-fluoro-5-nitrobenzyl bromide
4-Bromo-2-fluoro-5-nitrobenzyl bromide
Formel: C₇H₄Br₂FNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD17168700
CAS-Nummer: 1379295-27-8
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR