Potassium (2-fluoro-5-formylphenyl)trifluoroborate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium trifluoro(2-fluoro-5-formylphenyl)borate
PC53342-5G
PC53342-25G
APOSPC53342-5GEA
69.6
EUR
APOSPC53342-5G
APOSPC53342-25G
Potassium (2-fluoro-5-formylphenyl)trifluoroborate 95%
Potassium (2-fluoro-5-formylphenyl)trifluoroborate
Formel:
C₇H₄BF₄O·K Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09993332 CAS-Nummer: 1012868-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...