Methyl 3-bromo-2-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC53202-1G PC53202-250MG PC53202-5G
APOSPC53202-1GEA 364 EUR
APOSPC53202-1G APOSPC53202-250MG APOSPC53202-5G
Methyl 3-bromo-2-(trifluoromethyl)benzoate
Methyl 3-bromo-2-(trifluoromethyl)benzoate
Formel: C₉H₆BrF₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD14697997
CAS-Nummer: 1214323-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR