3-Ethyl-4-fluorobromobenzene 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-ethyl-1-fluorobenzene
PC53137-1G
PC53137-5G
APOSPC53137-1GEA
36.4
EUR
APOSPC53137-1G
APOSPC53137-5G
3-Ethyl-4-fluorobromobenzene 98%
3-Ethyl-4-fluorobromobenzene
Formel:
C₈H₈BrF Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11846037 CAS-Nummer: 627463-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...