Methyl 3-bromo-2-chloro-6-fluorobenzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53024-1G
PC53024-5G
APOSPC53024-1GEA
65.4
EUR
APOSPC53024-1G
APOSPC53024-5G
Methyl 3-bromo-2-chloro-6-fluorobenzoate 95%
Methyl 3-bromo-2-chloro-6-fluorobenzoate
Formel:
C₈H₅BrClFO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28140551 CAS-Nummer: 1784053-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...