4-Fluoro-DL-homophenylalanine 97%
Lieferant: Apollo Scientific
PC530008-1G
PC530008-250MG
PC530008-500MG
APOSPC530008-1GEA
654
EUR
APOSPC530008-1G
APOSPC530008-250MG
APOSPC530008-500MG
4-Fluoro-DL-homophenylalanine 97%
4-Fluoro-DL-homophenylalanine
Formel:
C₁₀H₁₂FNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07372156 CAS-Nummer: 225233-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...