2,4-Difluoro-D-phenylalanine 97%
Lieferant: Apollo Scientific
Synonyme:
(R)-2-Amino-3-(2,4-difluorophenyl)propanoic acid
PC530002-1G
PC530002-250MG
APOSPC530002-1GEA
213
EUR
APOSPC530002-1G
APOSPC530002-250MG
2,4-Difluoro-D-phenylalanine 97%
2,4-Difluoro-D-phenylalanine
Formel:
C₉H₉F₂NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07371994 CAS-Nummer: 266360-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...