4-Bromo-2-fluoro-1-phenoxybenzene 98%
Lieferant: Apollo Scientific
Synonyme:
1-Bromo-3-fluoro-4-phenoxybenzene
PC52476-1G
PC52476-5G
APOSPC52476-1GEA
132
EUR
APOSPC52476-1G
APOSPC52476-5G
4-Bromo-2-fluoro-1-phenoxybenzene 98%
4-Bromo-2-fluoro-1-phenoxybenzene
Formel:
C₁₂H₈BrFO Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
230961-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...