2-Amino-7-fluoroindole hydrochloride

Lieferant: Apollo Scientific

PC52235-1G PC52235-250MG
APOSPC52235-1GEA 654 EUR
APOSPC52235-1G APOSPC52235-250MG
2-Amino-7-fluoroindole hydrochloride
Formel: C₈H₇FN₂·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD32638671
CAS-Nummer: 365548-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR