2-Amino-6-fluoroindole hydrochloride 95%

Lieferant: Apollo Scientific

PC52234-1G PC52234-250MG
APOSPC52234-1GEA 654 EUR
APOSPC52234-1G APOSPC52234-250MG
2-Amino-6-fluoroindole hydrochloride 95%
2-Amino-6-fluoroindole hydrochloride
Formel: C₈H₇FN₂·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20727221
CAS-Nummer: 1262587-80-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR