4-Bromo-2-chloro-3-fluorobenzyl bromide 95%
Lieferant: Apollo Scientific
PC52204-1G
PC52204-250MG
APOSPC52204-1GEA
579
EUR
APOSPC52204-1G
APOSPC52204-250MG
4-Bromo-2-chloro-3-fluorobenzyl bromide 95%
4-Bromo-2-chloro-3-fluorobenzyl bromide
Formel:
C₇H₄Br₂ClF Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
2090530-74-6 UN-Nummer: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...