4-(Fluoromethyl)aniline, N-BOC protected 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-Butyl [4-(fluoromethyl)phenyl]carbamate

PC52152-1G PC52152-250MG
APOSPC52152-1GEA 654 EUR
APOSPC52152-1G APOSPC52152-250MG
4-(Fluoromethyl)aniline, N-BOC protected 95%
4-(Fluoromethyl)aniline, N-BOC protected
Formel: C₁₂H₁₆FNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31812098
CAS-Nummer: 2366994-54-7
UN-Nummer: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR