1-(Trifluoromethyl)naphthalene 97%

Lieferant: Apollo Scientific

PC520796-10G PC520796-1G PC520796-5G
APOSPC520796-10GEA 882 EUR
APOSPC520796-10G APOSPC520796-1G APOSPC520796-5G
1-(Trifluoromethyl)naphthalene 97%
1-(Trifluoromethyl)naphthalene
Formel: C₁₁H₇F₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04973005
CAS-Nummer: 26458-04-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR