4-(1-Fluoro-1-methyl-ethyl)-1H-pyrimidin-6-one 95%

Lieferant: Apollo Scientific

PC520782-1G PC520782-5G
APOSPC520782-1GEA 268 EUR
APOSPC520782-1G APOSPC520782-5G
4-(1-Fluoro-1-methyl-ethyl)-1H-pyrimidin-6-one 95%
4-(1-Fluoro-1-methyl-ethyl)-1H-pyrimidin-6-one
Formel: C₇H₉FN₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD24604646
CAS-Nummer: 240414-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR