3-Bromo-5-fluoro-pyridine-4-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-5-fluoroisonicotinic acid, 3-Bromo-5-fluoropyridine-4-carboxylic acid
PC520692-1G
PC520692-5G
APOSPC520692-1GEA
58
EUR
APOSPC520692-1G
APOSPC520692-5G
3-Bromo-5-fluoro-pyridine-4-carboxylic acid
3-Bromo-5-fluoro-pyridine-4-carboxylic acid
Formel:
C₆H₃BrFNO₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD13181633 CAS-Nummer: 955372-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...