2,4-Difluoro-3-methylbenzyl alcohol 97%
Lieferant: Apollo Scientific
PC520556-1G
PC520556-5G
APOSPC520556-1GEA
257
EUR
APOSPC520556-1G
APOSPC520556-5G
2,4-Difluoro-3-methylbenzyl alcohol 97%
2,4-Difluoro-3-methylbenzyl alcohol
Formel:
C₈H₈F₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07777159 CAS-Nummer: 847502-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...