1-Chloro-3,5-difluoro-2-iodobenzene 95%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-2,4-difluoroiodobenzene
PC51904-1G
PC51904-250MG
PC51904-5G
APOSPC51904-1GEA
415
EUR
APOSPC51904-1G
APOSPC51904-250MG
APOSPC51904-5G
1-Chloro-3,5-difluoro-2-iodobenzene 95%
1-Chloro-3,5-difluoro-2-iodobenzene
Formel:
C₆H₂ClF₂I Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16817642 CAS-Nummer: 1242339-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...