2-Bromo-3-fluorophenylacetylene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-ethynyl-3-fluorobenzene
PC51222-1G
PC51222-250MG
APOSPC51222-1GEA
609
EUR
APOSPC51222-1G
APOSPC51222-250MG
2-Bromo-3-fluorophenylacetylene
2-Bromo-3-fluorophenylacetylene
Formel:
C₈H₄BrF Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD28127154 CAS-Nummer: 1855543-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...