2-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Carboxy-4-chloro-5-nitrobenzotrifluoride, 6-Chloro-5-nitro-α,α,α-trifluoro-m-toluic acid
PC51195-1G
PC51195-5G
APOSPC51195-1GEA
65.4
EUR
APOSPC51195-1G
APOSPC51195-5G
2-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
2-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₃ClF₃NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11847295 CAS-Nummer: 22227-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...