2-Bromo-5-fluoro-4-nitrotoluene 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-fluoro-4-methylnitrobenzene, 1-Bromo-4-fluoro-2-methyl-5-nitrobenzene

PC51134-1G PC51134-5G PC51134-25G
APOSPC51134-1GEA 45.1 EUR
APOSPC51134-1G APOSPC51134-5G APOSPC51134-25G
2-Bromo-5-fluoro-4-nitrotoluene 97%
2-Bromo-5-fluoro-4-nitrotoluene
Formel: C₇H₅BrFNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11110550
CAS-Nummer: 170098-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR