N-Methyl-2-nitro-3,4,5-trifluoroaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Methylamino)-2,3,4-trifluoronitrobenzene

PC51047-1G PC51047-5G
APOSPC51047-1GEA 207 EUR
APOSPC51047-1G APOSPC51047-5G
N-Methyl-2-nitro-3,4,5-trifluoroaniline
N-Methyl-2-nitro-3,4,5-trifluoroaniline
Formel: C₇H₅F₃N₂O₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD20481818
CAS-Nummer: 1346521-33-2
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR