4-Chloro-8-(trifluoromethyl)quinolin-2(1H)-one

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-1,2-dihydro-2-oxo-8-(trifluoromethyl)quinoline

PC51039-1G PC51039-5G
APOSPC51039-1GEA 177 EUR
APOSPC51039-1G APOSPC51039-5G
4-Chloro-8-(trifluoromethyl)quinolin-2(1H)-one
4-Chloro-8-(trifluoromethyl)quinolin-2(1H)-one
Formel: C₁₀H₅ClF₃NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05863554
CAS-Nummer: 955288-52-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR