Methyl 2,6-Difluoro-4-methylbenzoate
Lieferant: Apollo Scientific
PC510300-1G
PC510300-5G
APOSPC510300-1GEA
397
EUR
APOSPC510300-1G
APOSPC510300-5G
Methyl 2,6-Difluoro-4-methylbenzoate
Methyl 2,6-Difluoro-4-methylbenzoate
Formel:
C₉H₈F₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD23135649 CAS-Nummer: 79538-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...