Ethyl 2-(4,4-Difluorocyclohexyl)acetate

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-(4,4-difluorocyclohexyl)acetate

PC510248-1G PC510248-250MG
APOSPC510248-1GEA 865 EUR
APOSPC510248-1G APOSPC510248-250MG
Ethyl 2-(4,4-Difluorocyclohexyl)acetate
Ethyl 2-(4,4-Difluorocyclohexyl)acetate
Formel: C₁₀H₁₆F₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18909256
CAS-Nummer: 915213-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR