[6-(2-Fluorophenyl)-3-pyridyl]methanol

Lieferant: Apollo Scientific

PC510061-1G PC510061-5G
APOSPC510061-1GEA 499 EUR
APOSPC510061-1G APOSPC510061-5G
[6-(2-Fluorophenyl)-3-pyridyl]methanol
[6-(2-Fluorophenyl)-3-pyridyl]methanol
Formel: C₁₂H₁₀FNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06410429
CAS-Nummer: 887974-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR