2-[(Cyclopropylcarbonyl)amino]-4-fluorobenzoic acid

Lieferant: Apollo Scientific

PC510050-1G PC510050-5G
APOSPC510050-1GEA 275 EUR
APOSPC510050-1G APOSPC510050-5G
2-[(Cyclopropylcarbonyl)amino]-4-fluorobenzoic acid
2-[(Cyclopropylcarbonyl)amino]-4-fluorobenzoic acid
Formel: C₁₁H₁₀FNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09730560
CAS-Nummer: 953753-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR