N-Succinimidyl pentafluorobenzoate, technische Qualität
Lieferant: Apollo Scientific
PC50955-25G
PC50955-5G
APOSPC50955-25GEA
447
EUR
APOSPC50955-25G
APOSPC50955-5G
N-Succinimidyl pentafluorobenzoate, technische Qualität
N-Succinimidyl pentafluorobenzoate
Formel:
C₁₁H₄F₅NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD30532056 CAS-Nummer: 178942-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...