N-Methyl-pentafluorobenzamide

Lieferant: Apollo Scientific

PC50954-25G PC50954-5G
APOSPC50954-25GEA 241 EUR
APOSPC50954-25G APOSPC50954-5G
N-Methyl-pentafluorobenzamide
N-Methyl-pentafluorobenzamide
Formel: C₈H₄F₅NO
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 120033-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR