Methyl 2-fluoro-4-methoxybenzoate

Lieferant: Apollo Scientific

PC502590-1G PC502590-5G
APOSPC502590-1GEA 69.6 EUR
APOSPC502590-1G APOSPC502590-5G
Methyl 2-fluoro-4-methoxybenzoate
Methyl 2-fluoro-4-methoxybenzoate
Formel: C₉H₉FO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06203758
CAS-Nummer: 128272-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR