2,3-Difluoro-4-(trifluoromethoxy)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC502428-1G PC502428-250MG
APOSPC502428-1GEA 623 EUR
APOSPC502428-1G APOSPC502428-250MG
2,3-Difluoro-4-(trifluoromethoxy)benzoic acid 95%
2,3-Difluoro-4-(trifluoromethoxy)benzoic acid
Formel: C₈H₃F₅O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31561151
CAS-Nummer: 1309597-49-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR