3-Iodo-5-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC502144-1G PC502144-250MG
APOSPC502144-1GEA 1400 EUR
APOSPC502144-1G APOSPC502144-250MG
3-Iodo-5-(trifluoromethyl)benzyl alcohol
3-Iodo-5-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆F₃IO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18398176
CAS-Nummer: 868167-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR