2,4-Difluoro-3-iodobenzotrifluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Difluoro-2-iodo-4-(trifluoromethyl)benzene

PC502070-1G PC502070-250MG PC502070-5G
APOSPC502070-1GEA 480 EUR
APOSPC502070-1G APOSPC502070-250MG APOSPC502070-5G
2,4-Difluoro-3-iodobenzotrifluoride
2,4-Difluoro-3-iodobenzotrifluoride
Formel: C₇H₂F₅I
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD30481111
CAS-Nummer: 2089257-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR