2-Fluoro-5-iodonitrobenzene 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Fluoro-4-iodo-2-nitrobenzene
PC50206-1G
PC50206-5G
APOSPC50206-1GEA
21.9
EUR
APOSPC50206-1G
APOSPC50206-5G
2-Fluoro-5-iodonitrobenzene 97%
2-Fluoro-5-iodonitrobenzene
Formel:
C₆H₃FINO₂ Schmelzpunkt: 34…36 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09800783 CAS-Nummer: 364-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...