4-Bromo-5-fluoro-2-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

PC502009-1G PC502009-250MG
APOSPC502009-1GEA 248 EUR
APOSPC502009-1G APOSPC502009-250MG
4-Bromo-5-fluoro-2-methoxybenzoic acid
4-Bromo-5-fluoro-2-methoxybenzoic acid
Formel: C₈H₆BrFO₃
Molekulargewicht: 249.034 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD25459672
CAS-Nummer: 1780187-45-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR