3-Bromo-5-fluoro-2-nitroanisole

Lieferant: Apollo Scientific
Achtung

PC502008-1G PC502008-250MG PC502008-5G
APOSPC502008-1GEA 34.8 EUR
APOSPC502008-1G APOSPC502008-250MG APOSPC502008-5G
3-Bromo-5-fluoro-2-nitroanisole
3-Bromo-5-fluoro-2-nitroanisole
Formel: C₇H₅BrFNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28739839
CAS-Nummer: 1805593-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR