4’-Bromo-2’-fluoro-6’-methoxyacetophenone

Lieferant: Apollo Scientific
Gefahr

PC501761-1G PC501761-250MG
APOSPC501761-1GEA 754 EUR
APOSPC501761-1G APOSPC501761-250MG
4’-Bromo-2’-fluoro-6’-methoxyacetophenone
4’-Bromo-2’-fluoro-6’-methoxyacetophenone
Formel: C₉H₈BrFO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22570394
CAS-Nummer: 1822786-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR