4-Bromo-2-fluoro-6-methoxyaniline
Lieferant: Apollo Scientific
|
Achtung
|
PC501277-1G
PC501277-5G
PC501277-25G
APOSPC501277-1GEA
277
EUR
APOSPC501277-1G
APOSPC501277-5G
APOSPC501277-25G
4-Bromo-2-fluoro-6-methoxyaniline
4-Bromo-2-fluoro-6-methoxyaniline
Formel:
C₇H₇BrFNO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22418610 CAS-Nummer: 1261216-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...