5-Bromo-3-fluoro-2-nitroaniline

Lieferant: Apollo Scientific
Achtung

PC501267-1G PC501267-250MG
APOSPC501267-1GEA 470 EUR
APOSPC501267-1G APOSPC501267-250MG
5-Bromo-3-fluoro-2-nitroaniline
5-Bromo-3-fluoro-2-nitroaniline
Formel: C₆H₄BrFN₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD23702246
CAS-Nummer: 1193385-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR