2-Nitro-6-(trifluoromethyl)phenylacetonitrile

Lieferant: Apollo Scientific

PC501103-1G PC501103-250MG
APOSPC501103-1GEA 393 EUR
APOSPC501103-1G APOSPC501103-250MG
2-Nitro-6-(trifluoromethyl)phenylacetonitrile
2-Nitro-6-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₅F₃N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13185779
CAS-Nummer: 105003-99-4
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR