4-(2,2,2-Trifluoroethoxy)benzoic acid 95+%

Lieferant: Apollo Scientific

PC500851-1G PC500851-5G
APOSPC500851-1GEA 33 EUR
APOSPC500851-1G APOSPC500851-5G
4-(2,2,2-Trifluoroethoxy)benzoic acid 95+%
4-(2,2,2-Trifluoroethoxy)benzoic acid
Formel: C₉H₇F₃O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05669500
CAS-Nummer: 27914-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR