2-(2,2,2-Trifluoroethoxy)benzoic acid
Lieferant: Apollo Scientific
PC500832-1G
PC500832-5G
APOSPC500832-1GEA
286
EUR
APOSPC500832-1G
APOSPC500832-5G
2-(2,2,2-Trifluoroethoxy)benzoic acid
2-(2,2,2-Trifluoroethoxy)benzoic acid
Formel:
C₉H₇F₃O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08556302 CAS-Nummer: 35480-46-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...