5-Bromo-2-(trifluoromethyl)benzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Bromo-2-(trifluoromethyl)phenyl)methanol, [5-Bromo-2-(trifluoromethyl)phenyl]methanol
PC500758-1G
PC500758-250MG
APOSPC500758-1GEA
393
EUR
APOSPC500758-1G
APOSPC500758-250MG
5-Bromo-2-(trifluoromethyl)benzyl alcohol
5-Bromo-2-(trifluoromethyl)benzyl alcohol
Formel:
C₈H₆BrF₃O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13185398 CAS-Nummer: 1214349-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...