Caesium hexafluorophosphate
Lieferant: Apollo Scientific
Synonyme:
Cesium hexafluorophosphate
PC50069-1G
PC50069-25G
PC50069-5G
APOSPC50069-1GEA
127
EUR
APOSPC50069-1G
APOSPC50069-25G
APOSPC50069-5G
Caesium hexafluorophosphate
Caesium hexafluorophosphate
Formel:
CsPF₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00799707 CAS-Nummer: 16893-41-7 UN-Nummer: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...