2-Bromo-4-fluoro-5-(trifluoromethoxy)phenol

Lieferant: Apollo Scientific
Achtung

PC500561-250MG PC500561-100MG
APOSPC500561-250MGEA 237 EUR
APOSPC500561-250MG APOSPC500561-100MG
2-Bromo-4-fluoro-5-(trifluoromethoxy)phenol
2-Bromo-4-fluoro-5-(trifluoromethoxy)phenol
Formel: C₇H₃BrF₄O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28096578
CAS-Nummer: 1807165-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR