4'-Bromo-3'-(trifluoromethyl)acetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[4-Bromo-3-(trifluoromethyl)phenyl]ethanone

PC500495-1G PC500495-250MG PC500495-5G
APOSPC500495-1GEA 435 EUR
APOSPC500495-1G APOSPC500495-250MG APOSPC500495-5G
4'-Bromo-3'-(trifluoromethyl)acetophenone
4'-Bromo-3'-(trifluoromethyl)acetophenone
Formel: C₉H₆BrF₃O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD24148371
CAS-Nummer: 120077-70-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR