Methyl 3-bromo-6-chloro-2-fluorobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
PC500480-1G
PC500480-5G
PC500480-250MG
APOSPC500480-1GEA
198
EUR
APOSPC500480-1G
APOSPC500480-5G
APOSPC500480-250MG
Methyl 3-bromo-6-chloro-2-fluorobenzoate
Methyl 3-bromo-6-chloro-2-fluorobenzoate
Formel:
C₈H₅BrClFO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28040512 CAS-Nummer: 1543224-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...